Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505796.2 | XP_003505844.1 | 3 - 4 | 381 | 71 | |
XM_007646286.1 | XP_007644476.1 | 4 - 5 | 483 | 71 | ||
XM_007646287.1 | XP_007644477.1 | 3 - 4 | 396 | 71 | ||
XM_007646288.1 | XP_007644478.1 | 3 - 4 | 360 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Recommended Assay Probe spans exons |