Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640057.1 | XP_007638247.1 | 3 - 4 | 1459 | 74 | |
GenBank mRNA | JI888052.1 | - | 3 - 4 | 1459 | 74 | |
JP064852.1 | - | 3 - 4 | 1459 | 74 |