Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495840.1 | XP_003495888.1 | 6 - 7 | 1803 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Protease Probe spans exons |