Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498597.1 | XP_003498645.1 | 5 - 6 | 512 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |