Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640253.1 | XP_007638443.1 | 2 - 3 | 153 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |