Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506116.1 | XP_003506164.1 | 3 - 4 | 372 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |