Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynlt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507572.2 | XP_003507620.1 | 4 - 5 | 329 | 66 | |
GenBank mRNA | JP065874.1 | - | 4 - 5 | 311 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |