Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640280.1 | XP_007638470.1 | 4 - 5 | 1235 | 97 | |
XM_007640281.1 | XP_007638471.1 | 4 - 5 | 1192 | 97 | ||
XM_007640282.1 | XP_007638472.1 | 4 - 5 | 1192 | 97 | ||
XM_007640283.1 | XP_007638473.1 | 4 - 5 | 1232 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |