Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514821.2 | XP_003514869.1 | 2 - 3 | 567 | 87 | |
XM_007654875.1 | XP_007653065.1 | 2 - 3 | 530 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |