Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497006.1 | XP_003497054.1 | 4 - 5 | 1186 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Probe spans exons |