Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497006.1 | XP_003497054.1 | 6 - 7 | 1428 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Probe spans exons |