Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501839.2 | XP_003501887.1 | 3 - 4 | 694 | 106 | |
XM_003501840.2 | XP_003501888.1 | 3 - 4 | 517 | 106 | ||
XM_007642480.1 | XP_007640670.1 | 3 - 4 | 694 | 106 | ||
XM_007642481.1 | XP_007640671.1 | 3 - 4 | 694 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |