Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512902.2 | XP_003512950.1 | 3 - 4 | 327 | 71 | |
XM_007653293.1 | XP_007651483.1 | 3 - 4 | 331 | 71 | ||
XM_007653294.1 | XP_007651484.1 | 3 - 4 | 372 | 71 | ||
GenBank mRNA | JI887523.1 | - | 3 - 4 | 329 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |