Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640605.1 | XP_007638795.1 | 2 - 3 | 486 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Probe spans exons |