Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501103.2 | XP_003501151.1 | 1 - 2 | 185 | 96 | |
XM_007641796.1 | XP_007639986.1 | 1 - 2 | 185 | 96 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Probe spans exons |