Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641739.1 | XP_007639929.1 | 3 - 4 | 388 | 74 | |
XM_007641740.1 | XP_007639930.1 | 3 - 4 | 363 | 74 | ||
XM_007641741.1 | XP_007639931.1 | 3 - 4 | 342 | 74 | ||
GenBank mRNA | JI880524.1 | - | 4 - 5 | 457 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |