Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss11f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651098.1 | XP_007649288.1 | 6 - 7 | 747 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Probe spans exons |