Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501269.2 | XP_003501317.1 | 3 - 4 | 507 | 57 | |
GenBank mRNA | JI874658.1 | - | 3 - 4 | 499 | 57 | |
JP044806.1 | - | 3 - 4 | 500 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |