Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512317.1 | XP_003512365.1 | 4 - 5 | 678 | 67 | |
XM_003512318.1 | XP_003512366.1 | 4 - 5 | 678 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |