Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007609862.1 | XP_007608052.1 | 10 - 11 | 1653 | 54 | |
GenBank mRNA | JI874354.1 | - | 3 - 4 | 424 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |