Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654665.1 | XP_007652855.1 | 1 - 2 | 72 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |