Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499263.2 | XP_003499311.1 | 1 - 2 | 227 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Recommended Assay Probe spans exons |