Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499263.2 | XP_003499311.1 | 2 - 3 | 356 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis Probe spans exons |