Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505390.2 | XP_003505438.1 | 6 - 7 | 1253 | 54 | |
GenBank mRNA | JI895802.1 | - | 3 - 4 | 425 | 54 | |
JP058971.1 | - | 3 - 4 | 425 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |