Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514867.2 | XP_003514915.1 | 12 - 13 | 1120 | 111 | |
XM_007654905.1 | XP_007653095.1 | 12 - 13 | 1121 | 111 | ||
GenBank mRNA | JI894043.1 | - | 12 - 13 | 1113 | 111 | |
JP062973.1 | - | 12 - 13 | 1113 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |