Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653246.1 | XP_007651436.1 | 2 - 3 | 701 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |