Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502597.2 | XP_003502645.1 | 6 - 7 | 654 | 64 | |
GenBank mRNA | JI863000.1 | - | 4 - 5 | 339 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |