Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rilpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510262.2 | XP_003510310.1 | 1 - 2 | 582 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |