Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649599.1 | XP_007647789.1 | 5 - 6 | 1139 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Probe spans exons |