Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651232.1 | XP_007649422.1 | 4 - 5 | 474 | 113 | |
GenBank mRNA | JI862907.1 | - | 4 - 5 | 474 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |