Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513351.2 | XP_003513399.1 | 2 - 3 | 197 | 85 | |
GenBank mRNA | JP054972.1 | - | 1 | 219 | 85 |