Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646751.1 | XP_007644941.1 | 8 - 9 | 1086 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Ion Channel 3' Most Recommended Assay Probe spans exons |