Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lysmd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640151.1 | XP_007638341.1 | 2 - 3 | 601 | 78 | |
GenBank mRNA | JI878367.1 | - | 3 - 4 | 486 | 78 | |
JP039434.1 | - | 3 - 4 | 486 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |