Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxo6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642477.1 | XP_007640667.1 | 2 - 3 | 221 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis 3' Most Recommended Assay Probe spans exons |