Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514041.2 | XP_003514089.1 | 4 | 712 | 99 | |
GenBank mRNA | JI886190.1 | - | 4 | 687 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |