Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp27x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505114.2 | XP_003505162.2 | 1 | 462 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Both primers and probe map within a single exon |