Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tifa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654804.1 | XP_007652994.1 | 2 | 473 | 115 | |
XM_007654806.1 | XP_007652996.1 | 2 | 895 | 115 | ||
XM_007654807.1 | XP_007652997.1 | 2 | 204 | 115 | ||
XM_007654808.1 | XP_007652998.1 | 2 | 116 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |