Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbed2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007619019.1 | XP_007617209.1 | 2 | 319 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |