Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051309.1 | XP_017906798.1 | 11 - 12 | 5541 | 76 | |
XM_018051310.1 | XP_017906799.1 | 10 - 11 | 5370 | 76 | ||
XM_018051311.1 | XP_017906800.1 | 10 - 11 | 5142 | 76 | ||
XM_018051312.1 | XP_017906801.1 | 9 - 10 | 4971 | 76 | ||
XM_018051313.1 | XP_017906802.1 | 9 - 10 | 3300 | 76 | ||
XM_018051314.1 | XP_017906803.1 | 7 - 8 | 2730 | 76 | ||
XM_018051315.1 | XP_017906804.1 | 6 - 7 | 2262 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |