Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059897.1 | XP_017915386.1 | 1 - 2 | 413 | 107 | |
GenBank mRNA | JO576522.1 | - | 7 - 8 | 833 | 107 | |
JT707780.1 | - | 1 - 2 | 56 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |