Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676496.3 | XP_005676553.1 | 3 - 4 | 339 | 71 | |
XM_005676497.3 | XP_005676554.1 | 3 - 4 | 1171 | 71 | ||
XM_005676498.3 | XP_005676555.1 | 3 - 4 | 339 | 71 | ||
XM_013972530.2 | XP_013827984.1 | 3 - 4 | 376 | 71 | ||
XM_013972532.2 | XP_013827986.1 | 3 - 4 | 317 | 71 | ||
XM_013972544.2 | XP_013827998.1 | 4 - 5 | 456 | 71 | ||
XM_018059907.1 | XP_017915396.1 | 4 - 5 | 421 | 71 | ||
GenBank mRNA | JO595987.2 | - | 5 - 6 | 2614 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |