Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BECN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693865.3 | XP_005693922.1 | 4 - 5 | 358 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |