Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057731.1 | XP_017913220.1 | 4 - 5 | 888 | 71 | |
XM_018057732.1 | XP_017913221.1 | 3 - 4 | 805 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |