Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005697575.3 | XP_005697632.2 | 4 - 5 | 440 | 93 | |
XM_005697576.3 | XP_005697633.2 | 5 - 6 | 624 | 93 | ||
XM_005697577.3 | XP_005697634.2 | 5 - 6 | 611 | 93 | ||
XM_005697578.3 | XP_005697635.2 | 3 - 4 | 409 | 93 | ||
XM_005697579.3 | XP_005697636.2 | 5 - 6 | 562 | 93 | ||
XM_005697580.3 | XP_005697637.2 | 4 - 5 | 580 | 93 | ||
XM_005697581.3 | XP_005697638.2 | 4 - 5 | 453 | 93 | ||
XM_018040857.1 | XP_017896346.1 | 3 - 4 | 410 | 93 | ||
GenBank mRNA | JI861744.1 | - | 2 - 3 | 249 | 93 | |
JO620017.2 | - | 1 - 2 | 105 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |