Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013975552.2 | XP_013831006.2 | 10 - 11 | 1412 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons |