Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291973.1 | NP_001278902.1 | 1 - 2 | 354 | 76 | |
GenBank mRNA | KC236981.1 | - | 1 - 2 | 354 | 76 | |
KM596772.1 | - | 1 - 2 | 354 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |