Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRABP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046300.1 | XP_017901789.1 | 3 - 4 | 572 | 76 | |
GenBank mRNA | KA306830.1 | - | 1 - 2 | 448 | 76 | |
KF483979.1 | - | 3 - 4 | 361 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |