Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674869.3 | XP_005674926.1 | 5 - 6 | 477 | 101 | |
GenBank mRNA | JO341311.2 | - | 5 - 6 | 446 | 101 | |
JO362713.2 | - | 5 - 6 | 429 | 101 | ||
JO377262.2 | - | 5 - 6 | 439 | 101 | ||
JO421375.2 | - | 5 - 6 | 471 | 101 | ||
JO453403.2 | - | 1 - 2 | 138 | 101 | ||
JO539083.2 | - | 5 - 6 | 467 | 101 | ||
JO581235.1 | - | 5 - 6 | 511 | 101 | ||
JO632238.2 | - | 1 - 2 | 137 | 101 | ||
JR131732.1 | - | 4 - 5 | 357 | 101 | ||
JT668209.1 | - | 4 - 5 | 394 | 101 | ||
JT692850.1 | - | 4 - 5 | 395 | 101 | ||
JT730034.1 | - | 4 - 5 | 375 | 101 | ||
JT753906.1 | - | 4 - 5 | 393 | 101 | ||
KA343066.1 | - | 1 - 2 | 118 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |