Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698396.3 | XP_005698453.2 | 4 - 5 | 534 | 105 | |
GenBank mRNA | JI861751.1 | - | 4 - 5 | 468 | 105 | |
JO517302.2 | - | 1 - 2 | 188 | 105 | ||
JO537207.2 | - | 1 - 2 | 187 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |