Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699520.3 | XP_005699577.1 | 4 - 5 | 577 | 67 | |
XM_018043530.1 | XP_017899019.1 | 3 - 4 | 401 | 67 | ||
GenBank mRNA | JP350874.2 | - | 2 - 3 | 361 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |